missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Bis(3-aminophenoxy)diphenyl sulfone, 95%, Thermo Scientific™
$72.27 - $72.27
Chemical Identifiers
CAS | 30203-11-3 |
---|---|
Molecular Formula | C24H20N2O4S |
Molecular Weight (g/mol) | 432.49 |
MDL Number | MFCD00054738 |
InChI Key | WCXGOVYROJJXHA-UHFFFAOYSA-N |
Synonym | 3,3'-sulfonylbis 4,1-phenylene bis oxy dianiline, bis 4-3-aminophenoxy phenyl sulfone, 4,4'-bis 3-aminophenoxy diphenyl sulfone, benzenamine, 3,3'-sulfonylbis 4,1-phenyleneoxy bis, 4,4-bis 3-aminophenoxy diphenyl sulfone, 3-4-4-3-aminophenoxy benzenesulfonyl phenoxy aniline, 4-3-aminophenoxy-1-4-3-aminophenoxy phenyl sulfonyl benzene, acmc-209hds, 3,3'-sulfonylbis 4,1-phenyleneoxy dianiline |
PubChem CID | 121657 |
IUPAC Name | 3-[4-[4-(3-aminophenoxy)phenyl]sulfonylphenoxy]aniline |
SMILES | NC1=CC(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC(N)=CC=C3)C=C2)=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1719418
|
Thermo Scientific Chemicals
A1719418 |
50 g |
Each for $72.27
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
30203-11-3 | |
432.49 | |
WCXGOVYROJJXHA-UHFFFAOYSA-N | |
121657 | |
NC1=CC(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC(N)=CC=C3)C=C2)=CC=C1 |
C24H20N2O4S | |
MFCD00054738 | |
3,3'-sulfonylbis 4,1-phenylene bis oxy dianiline, bis 4-3-aminophenoxy phenyl sulfone, 4,4'-bis 3-aminophenoxy diphenyl sulfone, benzenamine, 3,3'-sulfonylbis 4,1-phenyleneoxy bis, 4,4-bis 3-aminophenoxy diphenyl sulfone, 3-4-4-3-aminophenoxy benzenesulfonyl phenoxy aniline, 4-3-aminophenoxy-1-4-3-aminophenoxy phenyl sulfonyl benzene, acmc-209hds, 3,3'-sulfonylbis 4,1-phenyleneoxy dianiline | |
3-[4-[4-(3-aminophenoxy)phenyl]sulfonylphenoxy]aniline |
Specifications
30203-11-3 | |
C24H20N2O4S | |
50 g | |
WCXGOVYROJJXHA-UHFFFAOYSA-N | |
3-[4-[4-(3-aminophenoxy)phenyl]sulfonylphenoxy]aniline | |
121657 | |
95% |
132°C to 135°C | |
MFCD00054738 | |
3,3'-sulfonylbis 4,1-phenylene bis oxy dianiline, bis 4-3-aminophenoxy phenyl sulfone, 4,4'-bis 3-aminophenoxy diphenyl sulfone, benzenamine, 3,3'-sulfonylbis 4,1-phenyleneoxy bis, 4,4-bis 3-aminophenoxy diphenyl sulfone, 3-4-4-3-aminophenoxy benzenesulfonyl phenoxy aniline, 4-3-aminophenoxy-1-4-3-aminophenoxy phenyl sulfonyl benzene, acmc-209hds, 3,3'-sulfonylbis 4,1-phenyleneoxy dianiline | |
NC1=CC(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC(N)=CC=C3)C=C2)=CC=C1 | |
432.49 | |
432.5 | |
4,4'-Bis(3-aminophenoxy)diphenyl sulfone |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 250-091-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only