missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Diaminodiphenylmethane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A156450B

 View more versions of this product

Catalog No. AAA156450B


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Diaminodiphenylmethane is used as an intermediate for the preparation of 4,4’-methylenediphenyldiisocyanate, 4,4′-methylenebis(cyclohexaneamine). It is also used in the preparation of polyurethane foams. Further, it is used as hardeners in epoxy resins and adhesives. In addition to this, it is used in the manufacturing of high-performance polymers.

Solubility
Soluble in alcohol, ether, acetone, benzene, ethyl ether, carbon tetrachloride and methanol. Slightly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, oxidizing acids, acid chlorides, acid anhydrides and chloroformates.
Specifications

Specifications

4,4'-Diaminodiphenylmethane
88°C to 92°C
221°C (429°F)
MFCD00007914
UN2651
14,2980
Soluble in alcohol,ether,acetone,benzene,ethyl ether,carbon tetrachloride and methanol. Slightly soluble in water.
NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
198.27
CHEBI:32506
97%
101-77-9
232°C (9 mmHg)
C13H14N2
1000 g
474706
4,4'-methylenedianiline, 4,4'-diaminodiphenylmethane, dadpm, methylenedianiline, dianilinomethane, tonox, p,p'-methylenedianiline, dianilinemethane, bis 4-aminophenyl methane, 4-4-aminobenzyl aniline
YBRVSVVVWCFQMG-UHFFFAOYSA-N
4-[(4-aminophenyl)methyl]aniline
7577
198.27

Chemical Identifiers

101-77-9
198.27
YBRVSVVVWCFQMG-UHFFFAOYSA-N
7577
4-[(4-aminophenyl)methyl]aniline
C13H14N2
MFCD00007914
4,4'-methylenedianiline, 4,4'-diaminodiphenylmethane, dadpm, methylenedianiline, dianilinomethane, tonox, p,p'-methylenedianiline, dianilinemethane, bis 4-aminophenyl methane, 4-4-aminobenzyl aniline
CHEBI:32506
NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H370-H341-H373-H302-H317
May cause cancer.
Causes damage to organs.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P260-P262-P264b-P270-P272-P280-P281-P301+P310-P302+P350-P305+P351+P338-P307+P311-P310-P330-P333+P313-P361-P363-P501c

H301-H310-H317-H319-H340-H350-H370-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 4,4'-DIAMINODIPHENYL METHANE

EINECSNumber : 202-974-4

RTECSNumber : BY5425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only