missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Diaminodiphenylmethane 98.0+%, TCI America™

Supplier:  TCI America M0220500G

 View more versions of this product

Catalog No. M0220500G


Explore available promotions

Description

Description

Specifications

Specifications

4,4′-Diaminodiphenylmethane
91°C
257°C
MFCD00007914
2651
YBRVSVVVWCFQMG-UHFFFAOYSA-N
4-[(4-aminophenyl)methyl]aniline
7577
198.27
Pellets
101-77-9
White-Yellow
C13H14N2
500 g
4,4'-methylenedianiline, 4,4'-diaminodiphenylmethane, dadpm, methylenedianiline, dianilinomethane, tonox, p,p'-methylenedianiline, dianilinemethane, bis 4-aminophenyl methane, 4-4-aminobenzyl aniline
NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
198.27
CHEBI:32506
≥98.0% (T)

Chemical Identifiers

101-77-9
198.27
YBRVSVVVWCFQMG-UHFFFAOYSA-N
7577
4-[(4-aminophenyl)methyl]aniline
C13H14N2
MFCD00007914
4,4'-methylenedianiline, 4,4'-diaminodiphenylmethane, dadpm, methylenedianiline, dianilinomethane, tonox, p,p'-methylenedianiline, dianilinemethane, bis 4-aminophenyl methane, 4-4-aminobenzyl aniline
CHEBI:32506
NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0040

RTECSNumber : BY5425000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions