missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Diaminodiphenylmethane 98.0+%, TCI America™

$32.87 - $76.13

Chemical Identifiers

CAS 101-77-9
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.27
MDL Number MFCD00007914
InChI Key YBRVSVVVWCFQMG-UHFFFAOYSA-N
Synonym 4,4'-methylenedianiline, 4,4'-diaminodiphenylmethane, dadpm, methylenedianiline, dianilinomethane, tonox, p,p'-methylenedianiline, dianilinemethane, bis 4-aminophenyl methane, 4-4-aminobenzyl aniline
PubChem CID 7577
ChEBI CHEBI:32506
IUPAC Name 4-[(4-aminophenyl)methyl]aniline
SMILES NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M022025G
View Documents
TCI America
M022025G
25 g
Each for $32.87
 
M0220500G
View Documents
TCI America
M0220500G
500 g
Each for $76.13
 
Description

Description

Specifications

Chemical Identifiers

101-77-9
198.27
YBRVSVVVWCFQMG-UHFFFAOYSA-N
7577
4-[(4-aminophenyl)methyl]aniline
C13H14N2
MFCD00007914
4,4'-methylenedianiline, 4,4'-diaminodiphenylmethane, dadpm, methylenedianiline, dianilinomethane, tonox, p,p'-methylenedianiline, dianilinemethane, bis 4-aminophenyl methane, 4-4-aminobenzyl aniline
CHEBI:32506
NC1=CC=C(CC2=CC=C(N)C=C2)C=C1

Specifications

101-77-9
White-Yellow
C13H14N2
25 g
4,4'-methylenedianiline, 4,4'-diaminodiphenylmethane, dadpm, methylenedianiline, dianilinomethane, tonox, p,p'-methylenedianiline, dianilinemethane, bis 4-aminophenyl methane, 4-4-aminobenzyl aniline
NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
198.27
CHEBI:32506
≥98.0% (T)
4,4′-Diaminodiphenylmethane
91°C
257°C
MFCD00007914
2651
YBRVSVVVWCFQMG-UHFFFAOYSA-N
4-[(4-aminophenyl)methyl]aniline
7577
198.27
Pellets
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0040

RTECSNumber : BY5425000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'