missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2783006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt | |
>300°C | |
MFCD00020350 | |
p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+] | |
291.286 | |
291.28 |
845-46-5 | |
C15H14N3NaO2 | |
5 g | |
OSCKRHPYZNTEIO-UHFFFAOYSA-M | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate | |
23674498 | |
Indicator |
Chemical Identifiers
845-46-5 | |
291.286 | |
OSCKRHPYZNTEIO-UHFFFAOYSA-M | |
23674498 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+] |
C15H14N3NaO2 | |
MFCD00020350 | |
p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only