missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals
$342.25 - $342.25
Chemical Identifiers
CAS | 845-46-5 |
---|---|
Molecular Formula | C15H14N3NaO2 |
Molecular Weight (g/mol) | 291.286 |
MDL Number | MFCD00020350 |
InChI Key | OSCKRHPYZNTEIO-UHFFFAOYSA-M |
Synonym | p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid |
PubChem CID | 23674498 |
IUPAC Name | sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2783006
|
Thermo Scientific Chemicals
H2783006 |
5 g |
Each for $342.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
845-46-5 | |
291.286 | |
OSCKRHPYZNTEIO-UHFFFAOYSA-M | |
23674498 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+] |
C15H14N3NaO2 | |
MFCD00020350 | |
p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
Specifications
845-46-5 | |
C15H14N3NaO2 | |
5 g | |
OSCKRHPYZNTEIO-UHFFFAOYSA-M | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate | |
23674498 | |
Indicator |
>300°C | |
MFCD00020350 | |
p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+] | |
291.286 | |
291.28 | |
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only