missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetophenetidide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 216595000

 View more versions of this product

Catalog No. AC216595000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Acetophenetidide
62-44-2
White
96% min. (HPLC)
C10H13NO2
MFCD00009094
13, 461
phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin
CPJSUEIXXCENMM-UHFFFAOYSA-N
N-(4-ethoxyphenyl)acetamide
4754
179.22
Fine Crystalline Powder
97%
133.0°C to 138.0°C
Authentic
Plastic bottle
CH3CONHC6H4OC2H5
500 g
15, 7319
Solubility in water: 1g/1310mL cold water. Other solubilities: 1g/82mL boiling water, 1g/15mL cold alcohol, 1g/2.8mL boiling alcohol, 1g/14mL chloroform, 1g/90mL ether - soluble in glycerol
CCOC1=CC=C(C=C1)NC(=O)C
179.22
CHEBI:8050
97%

Chemical Identifiers

62-44-2
179.22
CPJSUEIXXCENMM-UHFFFAOYSA-N
4754
N-(4-ethoxyphenyl)acetamide
C10H13NO2
MFCD00009094
phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin
CHEBI:8050
CCOC1=CC=C(C=C1)NC(=O)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.
Harmful if swallowed.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 200-533-0

RTECSNumber : AM4375000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only