missing translation for 'onlineSavingsMsg'
Learn More

4-Acetophenetidide, 97%

CAS: 62-44-2 | C10H13NO2 | 179.22 g/mol

$62.01 - $659.38

Chemical Identifiers

CAS 62-44-2
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00009094
InChI Key CPJSUEIXXCENMM-UHFFFAOYSA-N
Synonym phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin
PubChem CID 4754
ChEBI CHEBI:8050
IUPAC Name N-(4-ethoxyphenyl)acetamide
SMILES CCOC1=CC=C(C=C1)NC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC216591000
SDS View Documents
Thermo Scientific Chemicals
216591000
100 g
Each for $62.01
Only null left
 
AC216595000
SDS View Documents
Thermo Scientific Chemicals
216595000
500 g
Each for $189.15
Only null left
 
AC216590025
SDS View Documents
Thermo Scientific Chemicals
216590025
2.5 kg
Each for $659.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62-44-2
179.22
CPJSUEIXXCENMM-UHFFFAOYSA-N
4754
N-(4-ethoxyphenyl)acetamide
C10H13NO2
MFCD00009094
phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin
CHEBI:8050
CCOC1=CC=C(C=C1)NC(=O)C

Specifications

62-44-2
White
96% min. (HPLC)
C10H13NO2
MFCD00009094
13, 461
phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin
CPJSUEIXXCENMM-UHFFFAOYSA-N
N-(4-ethoxyphenyl)acetamide
4754
179.22
Fine Crystalline Powder
133.0°C to 138.0°C
Authentic
Plastic bottle
CH3CONHC6H4OC2H5
100 g
15, 7319
Solubility in water: 1g/1310mL cold water. Other solubilities: 1g/82mL boiling water, 1g/15mL cold alcohol, 1g/2.8mL boiling alcohol, 1g/14mL chloroform, 1g/90mL ether - soluble in glycerol
CCOC1=CC=C(C=C1)NC(=O)C
179.22
CHEBI:8050
97%
4-Acetophenetidide, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.
Harmful if swallowed.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 200-533-0

RTECSNumber : AM4375000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only