Learn More
4-Allyl-1,6-heptadien-4-ol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2070388
Description
A highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
4-Allyl-1,6-heptadien-4-ol | |
C10H16O | |
2.5 g | |
Soluble in water. | |
C=CCC(CC=C)(CC=C)O | |
152.237 | |
152.24 |
10202-75-2 | |
MFCD00039829 | |
4-allyl-1,6-heptadien-4-ol, triallylcarbinol, 4-allylhepta-1,6-dien-4-ol, 4-allyl-1,6-heptadiene-4-ol, 1,6-heptadien-4-ol, 4-2-propenyl, 4-prop-2-en-1-yl hepta-1,6-dien-4-ol, triallyl carbinol, acmc-1byg9, 4-allyl-hepta-1,6-dien-4-ol, 1,6-heptadien-4-ol,4-2-propen-1-yl | |
SUXQWOWVXDXQSE-UHFFFAOYSA-N | |
4-prop-2-enylhepta-1,6-dien-4-ol | |
139107 | |
99% |
Chemical Identifiers
10202-75-2 | |
152.237 | |
SUXQWOWVXDXQSE-UHFFFAOYSA-N | |
139107 | |
C=CCC(CC=C)(CC=C)O |
C10H16O | |
MFCD00039829 | |
4-allyl-1,6-heptadien-4-ol, triallylcarbinol, 4-allylhepta-1,6-dien-4-ol, 4-allyl-1,6-heptadiene-4-ol, 1,6-heptadien-4-ol, 4-2-propenyl, 4-prop-2-en-1-yl hepta-1,6-dien-4-ol, triallyl carbinol, acmc-1byg9, 4-allyl-hepta-1,6-dien-4-ol, 1,6-heptadien-4-ol,4-2-propen-1-yl | |
4-prop-2-enylhepta-1,6-dien-4-ol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only