Learn More
4-Allyl-1,6-heptadien-4-ol, 99%, Thermo Scientific Chemicals
$63.18 - $684.33
Chemical Identifiers
CAS | 10202-75-2 |
---|---|
Molecular Formula | C10H16O |
Molecular Weight (g/mol) | 152.237 |
MDL Number | MFCD00039829 |
InChI Key | SUXQWOWVXDXQSE-UHFFFAOYSA-N |
Synonym | 4-allyl-1,6-heptadien-4-ol, triallylcarbinol, 4-allylhepta-1,6-dien-4-ol, 4-allyl-1,6-heptadiene-4-ol, 1,6-heptadien-4-ol, 4-2-propenyl, 4-prop-2-en-1-yl hepta-1,6-dien-4-ol, triallyl carbinol, acmc-1byg9, 4-allyl-hepta-1,6-dien-4-ol, 1,6-heptadien-4-ol,4-2-propen-1-yl |
PubChem CID | 139107 |
IUPAC Name | 4-prop-2-enylhepta-1,6-dien-4-ol |
SMILES | C=CCC(CC=C)(CC=C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2070388
|
Thermo Scientific Chemicals
B2070388 |
2.5 g |
Each for $63.18
|
|
|||||
AAB2070309
|
Thermo Scientific Chemicals
B2070309 |
10 g |
Each for $166.45
|
|
|||||
AAB2070318
|
Thermo Scientific Chemicals
B2070318 |
50 g |
Each for $684.33
|
|
|||||
Description
A highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
10202-75-2 | |
152.237 | |
SUXQWOWVXDXQSE-UHFFFAOYSA-N | |
139107 | |
C=CCC(CC=C)(CC=C)O |
C10H16O | |
MFCD00039829 | |
4-allyl-1,6-heptadien-4-ol, triallylcarbinol, 4-allylhepta-1,6-dien-4-ol, 4-allyl-1,6-heptadiene-4-ol, 1,6-heptadien-4-ol, 4-2-propenyl, 4-prop-2-en-1-yl hepta-1,6-dien-4-ol, triallyl carbinol, acmc-1byg9, 4-allyl-hepta-1,6-dien-4-ol, 1,6-heptadien-4-ol,4-2-propen-1-yl | |
4-prop-2-enylhepta-1,6-dien-4-ol |
Specifications
10202-75-2 | |
MFCD00039829 | |
4-allyl-1,6-heptadien-4-ol, triallylcarbinol, 4-allylhepta-1,6-dien-4-ol, 4-allyl-1,6-heptadiene-4-ol, 1,6-heptadien-4-ol, 4-2-propenyl, 4-prop-2-en-1-yl hepta-1,6-dien-4-ol, triallyl carbinol, acmc-1byg9, 4-allyl-hepta-1,6-dien-4-ol, 1,6-heptadien-4-ol,4-2-propen-1-yl | |
SUXQWOWVXDXQSE-UHFFFAOYSA-N | |
4-prop-2-enylhepta-1,6-dien-4-ol | |
139107 | |
99% |
C10H16O | |
2.5 g | |
Soluble in water. | |
C=CCC(CC=C)(CC=C)O | |
152.237 | |
152.24 | |
4-Allyl-1,6-heptadien-4-ol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only