missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6273303
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesStore in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester | |
White | |
MFCD16996254 | |
3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-amino-2-chloro-3-fluorophenylboronic acid pinacol ester, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
CC1(C)OB(OC1(C)C)C1=C(Cl)C(F)=C(N)C=C1 | |
271.52 | |
271.52 | |
Solid |
1154740-65-4 | |
C12H16BClFNO2 | |
1 g | |
NFNKJOVOJSFUSA-UHFFFAOYSA-N | |
3-chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
59606745 | |
96% |
Chemical Identifiers
1154740-65-4 | |
271.52 | |
NFNKJOVOJSFUSA-UHFFFAOYSA-N | |
59606745 | |
CC1(C)OB(OC1(C)C)C1=C(Cl)C(F)=C(N)C=C1 |
C12H16BClFNO2 | |
MFCD16996254 | |
3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-amino-2-chloro-3-fluorophenylboronic acid pinacol ester, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
3-chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures