missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

$268.30 - $796.26

Chemical Identifiers

CAS 1154740-65-4
Molecular Formula C12H16BClFNO2
Molecular Weight (g/mol) 271.52
MDL Number MFCD16996254
InChI Key NFNKJOVOJSFUSA-UHFFFAOYSA-N
Synonym 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-amino-2-chloro-3-fluorophenylboronic acid pinacol ester, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 59606745
IUPAC Name 3-chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=C(Cl)C(F)=C(N)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62733MD
View Documents
Thermo Scientific Chemicals
H62733MD
250 mg
Each for $268.30
 
AAH6273303
View Documents
Thermo Scientific Chemicals
H6273303
1 g
Each for $796.26
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1154740-65-4
271.52
NFNKJOVOJSFUSA-UHFFFAOYSA-N
59606745
CC1(C)OB(OC1(C)C)C1=C(Cl)C(F)=C(N)C=C1
C12H16BClFNO2
MFCD16996254
3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-amino-2-chloro-3-fluorophenylboronic acid pinacol ester, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
3-chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

Specifications

1154740-65-4
C12H16BClFNO2
250 mg
NFNKJOVOJSFUSA-UHFFFAOYSA-N
3-chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
59606745
96%
4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester
White
MFCD16996254
3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-amino-2-chloro-3-fluorophenylboronic acid pinacol ester, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
CC1(C)OB(OC1(C)C)C1=C(Cl)C(F)=C(N)C=C1
271.52
271.52
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'