missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-nitrophenylboronic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 445300010

 View more versions of this product

Catalog No. AC445300010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Amino-3-nitrophenylboronic acid
96% min. (HPLC)
C6H7BN2O4
4-amino-3-nitrophenylboronic acid, 4-amino-3-nitrophenylboronicacid, 4-amino-3-nitrophenyl boronic acid, boronic acid,b-4-amino-3-nitrophenyl, pubchem4026, acmc-209r1c, irtxqnnjqznkrp-uhfffaoysa-n, 4-amino-3-nitro-phenyl boronic acid, 4-azanyl-3-nitro-phenyl boronic acid, boronic acid, b-4-amino-3-nitrophenyl
181.94
181.94
89466-07-9
Glass Bottle
1g
IRTXQNNJQZNKRP-UHFFFAOYSA-N
16217915
97%

Chemical Identifiers

89466-07-9
181.94
4-amino-3-nitrophenylboronic acid, 4-amino-3-nitrophenylboronicacid, 4-amino-3-nitrophenyl boronic acid, boronic acid,b-4-amino-3-nitrophenyl, pubchem4026, acmc-209r1c, irtxqnnjqznkrp-uhfffaoysa-n, 4-amino-3-nitro-phenyl boronic acid, 4-azanyl-3-nitro-phenyl boronic acid, boronic acid, b-4-amino-3-nitrophenyl
C6H7BN2O4
IRTXQNNJQZNKRP-UHFFFAOYSA-N
16217915
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only