missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrophenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 89466-07-9
Molecular Formula C6H7BN2O4
Molecular Weight (g/mol) 181.94
InChI Key IRTXQNNJQZNKRP-UHFFFAOYSA-N
Synonym 4-amino-3-nitrophenylboronic acid, 4-amino-3-nitrophenylboronicacid, 4-amino-3-nitrophenyl boronic acid, boronic acid,b-4-amino-3-nitrophenyl, pubchem4026, acmc-209r1c, irtxqnnjqznkrp-uhfffaoysa-n, 4-amino-3-nitro-phenyl boronic acid, 4-azanyl-3-nitro-phenyl boronic acid, boronic acid, b-4-amino-3-nitrophenyl
PubChem CID 16217915
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445300010
SDS View Documents
Thermo Scientific Chemicals
445300010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445300050
SDS View Documents
Thermo Scientific Chemicals
445300050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

89466-07-9
181.94
4-amino-3-nitrophenylboronic acid, 4-amino-3-nitrophenylboronicacid, 4-amino-3-nitrophenyl boronic acid, boronic acid,b-4-amino-3-nitrophenyl, pubchem4026, acmc-209r1c, irtxqnnjqznkrp-uhfffaoysa-n, 4-amino-3-nitro-phenyl boronic acid, 4-azanyl-3-nitro-phenyl boronic acid, boronic acid, b-4-amino-3-nitrophenyl
C6H7BN2O4
IRTXQNNJQZNKRP-UHFFFAOYSA-N
16217915

Specifications

89466-07-9
Glass Bottle
1g
IRTXQNNJQZNKRP-UHFFFAOYSA-N
16217915
97%
96% min. (HPLC)
C6H7BN2O4
4-amino-3-nitrophenylboronic acid, 4-amino-3-nitrophenylboronicacid, 4-amino-3-nitrophenyl boronic acid, boronic acid,b-4-amino-3-nitrophenyl, pubchem4026, acmc-209r1c, irtxqnnjqznkrp-uhfffaoysa-n, 4-amino-3-nitro-phenyl boronic acid, 4-azanyl-3-nitro-phenyl boronic acid, boronic acid, b-4-amino-3-nitrophenyl
181.94
181.94
4-Amino-3-nitrophenylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only