missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-p-terphenyl 97.0+%, TCI America™
Supplier: TCI America A134025G
Specifications
4-Amino-p-terphenyl | |
201°C | |
MFCD00191302 | |
ATGIXVUZFPZOHP-UHFFFAOYSA-N | |
4-(4-phenylphenyl)aniline | |
350072 | |
≥97.0% (GC) |
7293-45-0 | |
C18H15N | |
25 g | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)N | |
245.325 | |
245.33 | |
Crystalline Powder |
Chemical Identifiers
7293-45-0 | |
245.325 | |
ATGIXVUZFPZOHP-UHFFFAOYSA-N | |
4-(4-phenylphenyl)aniline |
C18H15N | |
MFCD00191302 | |
350072 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)N |
Safety and Handling
TSCA : No