missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-p-terphenyl 97.0+%, TCI America™

$142.21 - $1414.10

Chemical Identifiers

CAS 7293-45-0
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.325
MDL Number MFCD00191302
InChI Key ATGIXVUZFPZOHP-UHFFFAOYSA-N
PubChem CID 350072
IUPAC Name 4-(4-phenylphenyl)aniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A13401G
View Documents
TCI America
A13401G
1 g
Each for $142.21
 
A13405G
View Documents
TCI America
A13405G
5 g
Each for $426.60
 
A134025G
View Documents
TCI America
A134025G
25 g
Each for $1,414.10
 
Description

Description

Specifications

Chemical Identifiers

7293-45-0
245.325
ATGIXVUZFPZOHP-UHFFFAOYSA-N
4-(4-phenylphenyl)aniline
C18H15N
MFCD00191302
350072
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)N

Specifications

7293-45-0
C18H15N
1 g
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)N
245.325
245.33
Crystalline Powder
201°C
MFCD00191302
ATGIXVUZFPZOHP-UHFFFAOYSA-N
4-(4-phenylphenyl)aniline
350072
≥97.0% (GC)
4-Amino-p-terphenyl
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'