Learn More
4-Aminophenol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 104270051
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Aminophenol | |
96.0 | |
187.0°C to 191.0°C | |
284.0°C | |
Authentic | |
Plastic Drum | |
H2NC6H4OH | |
13,427 | |
p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p | |
PLIKAWJENQZMHA-UHFFFAOYSA-N | |
4-aminophenol | |
403 | |
109.13 | |
Crystalline Powder |
123-30-8 | |
100.0 | |
White to Cream | |
189°C | |
96% min. (GC) | |
C6H7NO | |
5 kg | |
15,458 | |
Solubility in water: 15g/L (20°C). Other solubilities: soluble in alcohol, practically insoluble in benzene and chloroform, 9.3% in ethyl methyl ketone (58.5°C), 4.5% in abs.ethanol (0°C) | |
C1=CC(=CC=C1N)O | |
109.13 | |
CHEBI:17602 | |
97% |
Chemical Identifiers
123-30-8 | |
109.13 | |
p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p | |
CHEBI:17602 | |
C1=CC(=CC=C1N)O |
C6H7NO | |
PLIKAWJENQZMHA-UHFFFAOYSA-N | |
403 | |
4-aminophenol |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 204-616-2
RUO – Research Use Only