missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminophenol, 97%, Thermo Scientific Chemicals

$42.53 - $850.92

Chemical Identifiers

CAS 123-30-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
InChI Key PLIKAWJENQZMHA-UHFFFAOYSA-N
Synonym p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
PubChem CID 403
ChEBI CHEBI:17602
IUPAC Name 4-aminophenol
SMILES C1=CC(=CC=C1N)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC104270050
SDS View Documents
Thermo Scientific Chemicals
104270050
5 g Glass bottle
Each for $42.53
 
AC104272500
SDS View Documents
Thermo Scientific Chemicals
104272500
250 g Glass bottle
Each for $68.87
 
AC104270010
SDS View Documents
Thermo Scientific Chemicals
104270010
1 kg Glass bottle
Each for $202.59
 
AC104270051
SDS View Documents
Thermo Scientific Chemicals
104270051
5 kg Plastic Drum
Each for $850.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-30-8
109.13
p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
CHEBI:17602
C1=CC(=CC=C1N)O
C6H7NO
PLIKAWJENQZMHA-UHFFFAOYSA-N
403
4-aminophenol

Specifications

123-30-8
100.0
White to Cream
189°C
96% min. (GC)
C6H7NO
5 g
15,458
Solubility in water: 15g/L (20°C). Other solubilities: soluble in alcohol, practically insoluble in benzene and chloroform, 9.3% in ethyl methyl ketone (58.5°C), 4.5% in abs.ethanol (0°C)
C1=CC(=CC=C1N)O
109.13
CHEBI:17602
97%
4-Aminophenol
96.0
187.0°C to 191.0°C
284.0°C
Authentic
Glass bottle
H2NC6H4OH
13,427
p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
PLIKAWJENQZMHA-UHFFFAOYSA-N
4-aminophenol
403
109.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 204-616-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only