Learn More
4-Aminosalicylic acid, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 65-49-6 |
---|---|
Molecular Formula | C7H7NO3 |
Molecular Weight (g/mol) | 153.14 |
MDL Number | MFCD00007789 |
InChI Key | WUBBRNOQWQTFEX-UHFFFAOYSA-N |
Synonym | 4-aminosalicylic acid, aminosalicylic acid, p-aminosalicylic acid, rezipas, pamisyl, parasalindon, aminopar, deapasil, ferrosan, apacil |
PubChem CID | 4649 |
ChEBI | CHEBI:27565 |
IUPAC Name | 4-amino-2-hydroxybenzoic acid |
SMILES | C1=CC(=C(C=C1N)O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC104620050
|
Thermo Scientific Chemicals
104620050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC104621000
|
Thermo Scientific Chemicals
104621000 |
100g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC104625000
|
Thermo Scientific Chemicals
104625000 |
500g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
65-49-6 | |
153.14 | |
WUBBRNOQWQTFEX-UHFFFAOYSA-N | |
4649 | |
4-amino-2-hydroxybenzoic acid |
C7H7NO3 | |
MFCD00007789 | |
4-aminosalicylic acid, aminosalicylic acid, p-aminosalicylic acid, rezipas, pamisyl, parasalindon, aminopar, deapasil, ferrosan, apacil | |
CHEBI:27565 | |
C1=CC(=C(C=C1N)O)C(=O)O |
Specifications
65-49-6 | |
100.0 | |
99% | |
C7H7NO3 | |
MFCD00007789 | |
14, 579 | |
4-aminosalicylic acid, aminosalicylic acid, p-aminosalicylic acid, rezipas, pamisyl, parasalindon, aminopar, deapasil, ferrosan, apacil | |
WUBBRNOQWQTFEX-UHFFFAOYSA-N | |
4-amino-2-hydroxybenzoic acid | |
4649 | |
153.14 | |
4-Aminosalicylic acid, 99% |
98.5 | |
Authentic | |
Glass bottle | |
H2NC6H3(OH)CO2H | |
5g | |
15, 473 | |
Solubility in water: 2g/L (20°C). Other solubilities: soluble in dilute hno3 and dilute naoh,aceton,1g/21mL alcohol,slightly soluble in ether,practically insoluble in benzene,chloroform,,carbontetrachloride | |
C1=CC(=C(C=C1N)O)C(=O)O | |
153.14 | |
CHEBI:27565 | |
99% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-613-5
RTECSNumber : VO1225000
TSCA : TSCA
RUO â Research Use Only