missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyloxyaniline hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 152291000

 View more versions of this product

Catalog No. AC152291000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Benzyloxyaniline hydrochloride
51388-20-6
100.0
Brown
98%
C13H13NO·HCl
MFCD00012995
13, 439
Solubility in water: slightly soluble
C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl
235.72
235.72
Crystalline Powder
98%
97.5
228°C
Authentic
Plastic bottle
C6H5CH2OC6H4NH2·HCl
100 g
4-benzyloxyaniline hydrochloride, 4-benzyloxy aniline hydrochloride, 4-benzyloxyaniline hcl, benzenamine, 4-phenylmethoxy-, hydrochloride, 4-phenylmethoxyaniline hydrochloride, aniline, p-benzyloxy-hydrochloride, 4-benzyloxy phenyl amine hydrochloride, 4-phenylmethoxy phenylamine, chloride, acmc-20amop, pubchem21343
KQBDLOVXZHOAJI-UHFFFAOYSA-N
4-phenylmethoxyaniline;hydrochloride
2723831
98%

Chemical Identifiers

51388-20-6
235.72
KQBDLOVXZHOAJI-UHFFFAOYSA-N
2723831
C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl
C13H13NO·HCl
MFCD00012995
4-benzyloxyaniline hydrochloride, 4-benzyloxy aniline hydrochloride, 4-benzyloxyaniline hcl, benzenamine, 4-phenylmethoxy-, hydrochloride, 4-phenylmethoxyaniline hydrochloride, aniline, p-benzyloxy-hydrochloride, 4-benzyloxy phenyl amine hydrochloride, 4-phenylmethoxy phenylamine, chloride, acmc-20amop, pubchem21343
4-phenylmethoxyaniline;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 257-170-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only