missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Benzyloxyphenoxyacetic acid, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0104718
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Benzyloxyphenoxyacetic acid | |
139°C to 140°C | |
MFCD00014361 | |
2469260 | |
VXMSXBVTUNOSLL-UHFFFAOYSA-N | |
2-(4-phenylmethoxyphenoxy)acetic acid | |
563696 | |
95% |
38559-92-1 | |
C15H14O4 | |
50 g | |
4-benzyloxyphenoxyacetic acid, 4-benzyloxy phenoxy acetic acid, 4-benzyloxy phenoxyacetic acid, 2-4-benzyloxy phenoxy acetic acid, 4-phenylmethoxy phenoxy acetic acid, acetic acid, 4-phenylmethoxy phenoxy, 2-4-phenylmethoxy phenoxy acetic acid, enamine_002147, acmc-1agp8 | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)OCC(=O)O | |
258.273 | |
258.28 |
Chemical Identifiers
38559-92-1 | |
258.273 | |
VXMSXBVTUNOSLL-UHFFFAOYSA-N | |
563696 | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)OCC(=O)O |
C15H14O4 | |
MFCD00014361 | |
4-benzyloxyphenoxyacetic acid, 4-benzyloxy phenoxy acetic acid, 4-benzyloxy phenoxyacetic acid, 2-4-benzyloxy phenoxy acetic acid, 4-phenylmethoxy phenoxy acetic acid, acetic acid, 4-phenylmethoxy phenoxy, 2-4-phenylmethoxy phenoxy acetic acid, enamine_002147, acmc-1agp8 | |
2-(4-phenylmethoxyphenoxy)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 254-000-2
TSCA : No
Recommended Storage : Ambient temperatures