missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Boc-amino)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2879906

 View more versions of this product

Catalog No. AAH2879906


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Boc-amino)benzeneboronic acid
∽200°C (decomposition)
MFCD02093054
4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
CC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O
237.06
237.06
380430-49-9
C11H16BNO4
5 g
UBVOLHQIEQVXGM-UHFFFAOYSA-N
[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
3613184
97%

Chemical Identifiers

380430-49-9
237.06
UBVOLHQIEQVXGM-UHFFFAOYSA-N
3613184
CC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O
C11H16BNO4
MFCD02093054
4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only