missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Boc-amino)benzeneboronic acid, 97%, Thermo Scientific Chemicals

$71.05 - $237.40

Chemical Identifiers

CAS 380430-49-9
Molecular Formula C11H16BNO4
Molecular Weight (g/mol) 237.06
MDL Number MFCD02093054
InChI Key UBVOLHQIEQVXGM-UHFFFAOYSA-N
Synonym 4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
PubChem CID 3613184
IUPAC Name [4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2879903
View Documents
Thermo Scientific Chemicals
H2879903
1 g
Each for $71.05
 
AAH2879906
View Documents
Thermo Scientific Chemicals
H2879906
5 g
Each for $237.40
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

380430-49-9
237.06
UBVOLHQIEQVXGM-UHFFFAOYSA-N
3613184
CC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O
C11H16BNO4
MFCD02093054
4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid

Specifications

380430-49-9
C11H16BNO4
1 g
UBVOLHQIEQVXGM-UHFFFAOYSA-N
[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
3613184
97%
∽200°C (decomposition)
MFCD02093054
4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
CC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O
237.06
237.06
4-(Boc-amino)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only