missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-nitrophenylhydrazine hydrochloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2920203

 View more versions of this product

Catalog No. AAH2920203


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-2-nitrophenylhydrazine hydrochloride
∽185°C (decomposition)
MFCD09260482
4-bromo-2-nitrophenylhydrazine hydrochloride, 4-bromo-2-nitrophenyl hydrazine hydrochloride, acmc-20amnd, 4-bromo-2-nitrophenyl hydrazine;hydrochloride, 4-bromo-2-nitrophenylhydrazinehydrochloride, 4-bromo-2-nitrophenyl hydrazine-hydrogen chloride 1/1
Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O
268.50
268.5
100032-79-9
C6H7BrClN3O2
1 g
SIBURLORIWZQNT-UHFFFAOYSA-N
(4-bromo-2-nitrophenyl)hydrazine;hydrochloride
44119379
95%

Chemical Identifiers

100032-79-9
268.50
SIBURLORIWZQNT-UHFFFAOYSA-N
44119379
Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O
C6H7BrClN3O2
MFCD09260482
4-bromo-2-nitrophenylhydrazine hydrochloride, 4-bromo-2-nitrophenyl hydrazine hydrochloride, acmc-20amnd, 4-bromo-2-nitrophenyl hydrazine;hydrochloride, 4-bromo-2-nitrophenylhydrazinehydrochloride, 4-bromo-2-nitrophenyl hydrazine-hydrogen chloride 1/1
(4-bromo-2-nitrophenyl)hydrazine;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only