missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-nitrophenylhydrazine hydrochloride, 95%, Thermo Scientific Chemicals

$217.32 - $829.08

Chemical Identifiers

CAS 100032-79-9
Molecular Formula C6H7BrClN3O2
Molecular Weight (g/mol) 268.50
MDL Number MFCD09260482
InChI Key SIBURLORIWZQNT-UHFFFAOYSA-N
Synonym 4-bromo-2-nitrophenylhydrazine hydrochloride, 4-bromo-2-nitrophenyl hydrazine hydrochloride, acmc-20amnd, 4-bromo-2-nitrophenyl hydrazine;hydrochloride, 4-bromo-2-nitrophenylhydrazinehydrochloride, 4-bromo-2-nitrophenyl hydrazine-hydrogen chloride 1/1
PubChem CID 44119379
IUPAC Name (4-bromo-2-nitrophenyl)hydrazine;hydrochloride
SMILES Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2920203
View Documents
Thermo Scientific Chemicals
H2920203
1 g
Each for $217.32
 
AAH2920206
View Documents
Thermo Scientific Chemicals
H2920206
5 g
Each for $829.08
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100032-79-9
268.50
SIBURLORIWZQNT-UHFFFAOYSA-N
44119379
Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O
C6H7BrClN3O2
MFCD09260482
4-bromo-2-nitrophenylhydrazine hydrochloride, 4-bromo-2-nitrophenyl hydrazine hydrochloride, acmc-20amnd, 4-bromo-2-nitrophenyl hydrazine;hydrochloride, 4-bromo-2-nitrophenylhydrazinehydrochloride, 4-bromo-2-nitrophenyl hydrazine-hydrogen chloride 1/1
(4-bromo-2-nitrophenyl)hydrazine;hydrochloride

Specifications

100032-79-9
C6H7BrClN3O2
1 g
SIBURLORIWZQNT-UHFFFAOYSA-N
(4-bromo-2-nitrophenyl)hydrazine;hydrochloride
44119379
95%
∽185°C (decomposition)
MFCD09260482
4-bromo-2-nitrophenylhydrazine hydrochloride, 4-bromo-2-nitrophenyl hydrazine hydrochloride, acmc-20amnd, 4-bromo-2-nitrophenyl hydrazine;hydrochloride, 4-bromo-2-nitrophenylhydrazinehydrochloride, 4-bromo-2-nitrophenyl hydrazine-hydrogen chloride 1/1
Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O
268.50
268.5
4-Bromo-2-nitrophenylhydrazine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only