missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3-chlorophenol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1380106

 View more versions of this product

Catalog No. AAL1380106


Explore available promotions

Description

Description

4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Bromo-3-chlorophenol
64°C to 68°C
C6H4BrClO
5 g
phenol, 4-bromo-3-chloro, 3-chloro-4-bromophenol, 2-chloro-4-hydroxybromobenzene, pubchem3597, acmc-1bvpq, 4-bromo-3-chloro-phenol, intermediates-zcf02051, ksc493i5n, 4-bromanyl-3-chloranyl-phenol
FQEYHIPPYOSPLF-UHFFFAOYSA-N
4-bromo-3-chlorophenol
7018182
98%
13631-21-5
Phenol-like
MFCD00070744
3236445
Slightly soluble in water.
C1=CC(=C(C=C1O)Cl)Br
207.451
207.46

Chemical Identifiers

13631-21-5
207.451
FQEYHIPPYOSPLF-UHFFFAOYSA-N
7018182
C1=CC(=C(C=C1O)Cl)Br
C6H4BrClO
MFCD00070744
phenol, 4-bromo-3-chloro, 3-chloro-4-bromophenol, 2-chloro-4-hydroxybromobenzene, pubchem3597, acmc-1bvpq, 4-bromo-3-chloro-phenol, intermediates-zcf02051, ksc493i5n, 4-bromanyl-3-chloranyl-phenol
4-bromo-3-chlorophenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only