missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-3-methylaniline, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 6933-10-4 |
---|---|
Molecular Formula | C7H8BrN |
Molecular Weight (g/mol) | 186.05 |
MDL Number | MFCD00007828 |
InChI Key | SVWWNEYBEFASMP-UHFFFAOYSA-N |
Synonym | methyl 2-aminoisonicotinate, 2-aminopyridine-4-carboxylic acid methyl ester, 2-aminoisonicotinic acid methyl ester, 4-methoxycarbonyl pyridin-2-amine, 4-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl 2-amino-4-pyridinecarboxylate, methyl-2-aminopyridine-4-carboxylate, 2-amino-4-pyridinecarboxylic acid methylester, 2-amino-4-pyridinecarboxylic acid methyl ester, methyl-2-aminoisonicotinat |
PubChem CID | 236984 |
IUPAC Name | methyl 2-aminopyridine-4-carboxylate |
SMILES | COC(=O)C1=CC(=NC=C1)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC167502500
|
Thermo Scientific Chemicals
167502500 |
250g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
6933-10-4 | |
186.05 | |
SVWWNEYBEFASMP-UHFFFAOYSA-N | |
236984 | |
COC(=O)C1=CC(=NC=C1)N |
C7H8BrN | |
MFCD00007828 | |
methyl 2-aminoisonicotinate, 2-aminopyridine-4-carboxylic acid methyl ester, 2-aminoisonicotinic acid methyl ester, 4-methoxycarbonyl pyridin-2-amine, 4-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl 2-amino-4-pyridinecarboxylate, methyl-2-aminopyridine-4-carboxylate, 2-amino-4-pyridinecarboxylic acid methylester, 2-amino-4-pyridinecarboxylic acid methyl ester, methyl-2-aminoisonicotinat | |
methyl 2-aminopyridine-4-carboxylate |
Specifications
6933-10-4 | |
100.0 | |
96% min. (GC) | |
BrC6H3(CH3)NH2 | |
250g | |
methyl 2-aminoisonicotinate, 2-aminopyridine-4-carboxylic acid methyl ester, 2-aminoisonicotinic acid methyl ester, 4-methoxycarbonyl pyridin-2-amine, 4-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl 2-amino-4-pyridinecarboxylate, methyl-2-aminopyridine-4-carboxylate, 2-amino-4-pyridinecarboxylic acid methylester, 2-amino-4-pyridinecarboxylic acid methyl ester, methyl-2-aminoisonicotinat | |
SVWWNEYBEFASMP-UHFFFAOYSA-N | |
methyl 2-aminopyridine-4-carboxylate | |
236984 | |
97% |
98.5 | |
240.0°C | |
C7H8BrN | |
MFCD00007828 | |
12, 873 | |
Solubility in water: insoluble | |
COC(=O)C1=CC(=NC=C1)N | |
186.05 | |
186.05 | |
4-Bromo-3-methylaniline, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 230-056-3
RUO â Research Use Only