missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-o-xylene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1473614

 View more versions of this product

Catalog No. AAA1473614


Explore available promotions

Description

Description

4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-Bromo-o-xylene
1.37
25 g
C8H9Br
2040354
Not miscible in water.
CC1=C(C=C(C=C1)Br)C
185.064
185.07
583-71-1
214°C to 215°C
1.556
MFCD00000086
4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo
QOGHRLGTXVMRLM-UHFFFAOYSA-N
4-bromo-1,2-dimethylbenzene
68504
97%

Chemical Identifiers

583-71-1
185.064
QOGHRLGTXVMRLM-UHFFFAOYSA-N
68504
CC1=C(C=C(C=C1)Br)C
C8H9Br
MFCD00000086
4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo
4-bromo-1,2-dimethylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 209-518-3

RTECSNumber : CY9020400

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only