Learn More
4-Bromo-o-xylene, 97%, Thermo Scientific Chemicals
$63.37 - $556.99
Chemical Identifiers
CAS | 583-71-1 |
---|---|
Molecular Formula | C8H9Br |
Molecular Weight (g/mol) | 185.064 |
MDL Number | MFCD00000086 |
InChI Key | QOGHRLGTXVMRLM-UHFFFAOYSA-N |
Synonym | 4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo |
PubChem CID | 68504 |
IUPAC Name | 4-bromo-1,2-dimethylbenzene |
SMILES | CC1=C(C=C(C=C1)Br)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1473606
|
Thermo Scientific Chemicals
A1473606 |
5 g |
Each for $63.37
|
|
|||||
AAA1473614
|
Thermo Scientific Chemicals
A1473614 |
25 g |
Each for $188.10
|
|
|||||
AAA1473622
|
Thermo Scientific Chemicals
A1473622 |
100 g |
Each for $556.99
|
|
|||||
Description
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
583-71-1 | |
185.064 | |
QOGHRLGTXVMRLM-UHFFFAOYSA-N | |
68504 | |
CC1=C(C=C(C=C1)Br)C |
C8H9Br | |
MFCD00000086 | |
4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo | |
4-bromo-1,2-dimethylbenzene |
Specifications
583-71-1 | |
214°C to 215°C | |
1.556 | |
MFCD00000086 | |
4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo | |
QOGHRLGTXVMRLM-UHFFFAOYSA-N | |
4-bromo-1,2-dimethylbenzene | |
68504 | |
97% |
1.37 | |
5 g | |
C8H9Br | |
2040354 | |
Not miscible in water. | |
CC1=C(C=C(C=C1)Br)C | |
185.064 | |
185.07 | |
4-Bromo-o-xylene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 209-518-3
RTECSNumber : CY9020400
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only