Learn More
4-Bromo-o-xylene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1473606
Description
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
4-Bromo-o-xylene | |
1.37 | |
5 g | |
C8H9Br | |
2040354 | |
Not miscible in water. | |
CC1=C(C=C(C=C1)Br)C | |
185.064 | |
185.07 |
583-71-1 | |
214°C to 215°C | |
1.556 | |
MFCD00000086 | |
4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo | |
QOGHRLGTXVMRLM-UHFFFAOYSA-N | |
4-bromo-1,2-dimethylbenzene | |
68504 | |
97% |
Chemical Identifiers
583-71-1 | |
185.064 | |
QOGHRLGTXVMRLM-UHFFFAOYSA-N | |
68504 | |
CC1=C(C=C(C=C1)Br)C |
C8H9Br | |
MFCD00000086 | |
4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo | |
4-bromo-1,2-dimethylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 209-518-3
RTECSNumber : CY9020400
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only