missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromoaniline, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 146280250

 View more versions of this product

Catalog No. AC146280250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromoaniline
60.0°C to 64.0°C
1.4970g/mL
>110°C
99% min. (GC)
C6H6BrN
25 g
1.497
p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
WDFQBORIUYODSI-UHFFFAOYSA-N
4-bromoaniline
7807
99+%
106-40-1
Brown-Gray to White
230.0°C to 250.0°C
Authentic
Glass bottle
BrC6H4NH2
12,636
15,1411
Solubility in water: < 1 g / l. Other solubilities: soluble in most organic solvents except cold,petroleum,very soluble in alcohol and ether
C1=CC(=CC=C1N)Br
172.02
172.02
Crystalline Powder

Chemical Identifiers

106-40-1
172.02
p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
4-bromoaniline
C6H6BrN
WDFQBORIUYODSI-UHFFFAOYSA-N
7807
C1=CC(=CC=C1N)Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 203-393-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only