missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1148606

 View more versions of this product

Catalog No. AAL1148606


Explore available promotions

Description

Description

4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong bases and strong oxidizing agents.
Specifications

Specifications

4-Bromobenzyl chloride
38°C to 41°C
>110°C (230°F)
MFCD00040867
UN3261
14,1410
Insoluble in water.
ClCC1=CC=C(Br)C=C1
205.48
205.49
589-17-3
106°C to 107°C (10 mmHg)
C7H6BrCl
5 g
6123220
1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride
BSIIGUGKOPPTPZ-UHFFFAOYSA-N
1-bromo-4-(chloromethyl)benzene
68528
97%

Chemical Identifiers

589-17-3
205.48
BSIIGUGKOPPTPZ-UHFFFAOYSA-N
68528
ClCC1=CC=C(Br)C=C1
C7H6BrCl
MFCD00040867
1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride
1-bromo-4-(chloromethyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 209-638-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only