Learn More
4-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals
$92.59 - $306.79
Chemical Identifiers
CAS | 589-17-3 |
---|---|
Molecular Formula | C7H6BrCl |
Molecular Weight (g/mol) | 205.48 |
MDL Number | MFCD00040867 |
InChI Key | BSIIGUGKOPPTPZ-UHFFFAOYSA-N |
Synonym | 1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride |
PubChem CID | 68528 |
IUPAC Name | 1-bromo-4-(chloromethyl)benzene |
SMILES | ClCC1=CC=C(Br)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1148606
|
Thermo Scientific Chemicals
L1148606 |
5 g |
Each for $92.59
|
|
||||||
AAL1148614
|
Thermo Scientific Chemicals
L1148614 |
25 g |
Each for $306.79
|
|
||||||
Description
4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong bases and strong oxidizing agents.
Chemical Identifiers
589-17-3 | |
205.48 | |
BSIIGUGKOPPTPZ-UHFFFAOYSA-N | |
68528 | |
ClCC1=CC=C(Br)C=C1 |
C7H6BrCl | |
MFCD00040867 | |
1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride | |
1-bromo-4-(chloromethyl)benzene |
Specifications
589-17-3 | |
106°C to 107°C (10 mmHg) | |
C7H6BrCl | |
5 g | |
6123220 | |
1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride | |
BSIIGUGKOPPTPZ-UHFFFAOYSA-N | |
1-bromo-4-(chloromethyl)benzene | |
68528 | |
97% |
38°C to 41°C | |
>110°C (230°F) | |
MFCD00040867 | |
UN3261 | |
14,1410 | |
Insoluble in water. | |
ClCC1=CC=C(Br)C=C1 | |
205.48 | |
205.49 | |
4-Bromobenzyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 209-638-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only