missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Bromobutyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2304009

 View more versions of this product

Catalog No. AAB2304009


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(4-Bromobutyl)triphenylphosphonium bromide
206°C to 212°C
MFCD00031636
3812291
4-bromobutyl triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphonium bromide, 4-bromobutyl triphenylphosphanium bromide, 4-bromobutyl-triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphanium bromide, 4-bromobutyl triphenyl-phosphonium bromide, 4-bromo butyl triphenylphosphonium bromide, 4-bromanylbutyl triphenyl phosphanium bromide, phosphonium, 4-bromobutyl triphenyl-,bromide 1:1, phosphonium, 4-bromobutyl triphenyl-, bromide 1:1
[Br-].BrCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
478.21
478.22
7333-63-3
C22H23Br2P
10 g
Hygroscopic
BJDNCJGRAMGIRU-UHFFFAOYSA-M
4-bromobutyl(triphenyl)phosphanium;bromide
2733851
98%

Chemical Identifiers

7333-63-3
478.21
BJDNCJGRAMGIRU-UHFFFAOYSA-M
2733851
[Br-].BrCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C22H23Br2P
MFCD00031636
4-bromobutyl triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphonium bromide, 4-bromobutyl triphenylphosphanium bromide, 4-bromobutyl-triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphanium bromide, 4-bromobutyl triphenyl-phosphonium bromide, 4-bromo butyl triphenylphosphonium bromide, 4-bromanylbutyl triphenyl phosphanium bromide, phosphonium, 4-bromobutyl triphenyl-,bromide 1:1, phosphonium, 4-bromobutyl triphenyl-, bromide 1:1
4-bromobutyl(triphenyl)phosphanium;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only