missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(4-Bromobutyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals
$71.77 - $215.49
Chemical Identifiers
CAS | 7333-63-3 |
---|---|
Molecular Formula | C22H23Br2P |
Molecular Weight (g/mol) | 478.21 |
MDL Number | MFCD00031636 |
InChI Key | BJDNCJGRAMGIRU-UHFFFAOYSA-M |
Synonym | 4-bromobutyl triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphonium bromide, 4-bromobutyl triphenylphosphanium bromide, 4-bromobutyl-triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphanium bromide, 4-bromobutyl triphenyl-phosphonium bromide, 4-bromo butyl triphenylphosphonium bromide, 4-bromanylbutyl triphenyl phosphanium bromide, phosphonium, 4-bromobutyl triphenyl-,bromide 1:1, phosphonium, 4-bromobutyl triphenyl-, bromide 1:1 |
PubChem CID | 2733851 |
IUPAC Name | 4-bromobutyl(triphenyl)phosphanium;bromide |
SMILES | [Br-].BrCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2304009
|
Thermo Scientific Chemicals
B2304009 |
10 g |
Each for $71.77
|
|
|||||
AAB2304018
|
Thermo Scientific Chemicals
B2304018 |
50 g |
Each for $215.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7333-63-3 | |
478.21 | |
BJDNCJGRAMGIRU-UHFFFAOYSA-M | |
2733851 | |
[Br-].BrCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C22H23Br2P | |
MFCD00031636 | |
4-bromobutyl triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphonium bromide, 4-bromobutyl triphenylphosphanium bromide, 4-bromobutyl-triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphanium bromide, 4-bromobutyl triphenyl-phosphonium bromide, 4-bromo butyl triphenylphosphonium bromide, 4-bromanylbutyl triphenyl phosphanium bromide, phosphonium, 4-bromobutyl triphenyl-,bromide 1:1, phosphonium, 4-bromobutyl triphenyl-, bromide 1:1 | |
4-bromobutyl(triphenyl)phosphanium;bromide |
Specifications
7333-63-3 | |
C22H23Br2P | |
10 g | |
Hygroscopic | |
BJDNCJGRAMGIRU-UHFFFAOYSA-M | |
4-bromobutyl(triphenyl)phosphanium;bromide | |
2733851 | |
98% |
206°C to 212°C | |
MFCD00031636 | |
3812291 | |
4-bromobutyl triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphonium bromide, 4-bromobutyl triphenylphosphanium bromide, 4-bromobutyl-triphenylphosphonium bromide, 4-bromobutyl triphenyl phosphanium bromide, 4-bromobutyl triphenyl-phosphonium bromide, 4-bromo butyl triphenylphosphonium bromide, 4-bromanylbutyl triphenyl phosphanium bromide, phosphonium, 4-bromobutyl triphenyl-,bromide 1:1, phosphonium, 4-bromobutyl triphenyl-, bromide 1:1 | |
[Br-].BrCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
478.21 | |
478.22 | |
(4-Bromobutyl)triphenylphosphonium bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only