missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Bromomethyl)phenylacetic acid, 97%, Thermo Scientific Chemicals

$72.51 - $199.05

Chemical Identifiers

CAS 13737-36-5
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00010632
InChI Key WCOCCXZFEJGHTC-UHFFFAOYSA-N
Synonym 4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
PubChem CID 4519056
IUPAC Name 2-[4-(bromomethyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2774403
View Documents
Thermo Scientific Chemicals
H2774403
1 g
Each for $72.51
 
AAH2774406
View Documents
Thermo Scientific Chemicals
H2774406
5 g
Each for $199.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13737-36-5
229.073
WCOCCXZFEJGHTC-UHFFFAOYSA-N
4519056
C1=CC(=CC=C1CC(=O)O)CBr
C9H9BrO2
MFCD00010632
4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
2-[4-(bromomethyl)phenyl]acetic acid

Specifications

13737-36-5
C9H9BrO2
1 g
4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
C1=CC(=CC=C1CC(=O)O)CBr
229.073
229.07
4-(Bromomethyl)phenylacetic acid
178°C to 180°C
MFCD00010632
UN3261
WCOCCXZFEJGHTC-UHFFFAOYSA-N
2-[4-(bromomethyl)phenyl]acetic acid
4519056
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H332-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only