missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-methoxyaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2833106

 View more versions of this product

Catalog No. AAH2833106


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chloro-3-methoxyaniline
78°C to 81°C
C7H8ClNO
5 g
3-methoxy-4-chloroaniline, 4-chloro-3-methoxyphenylamine, 4-chloro-m-anisidine, 2-chloro-5-aminoanisole, 5-amino-2-chloroanisole, 4-chloro-3-methoxy aniline, 4-chloro-3-methoxy-phenylamine, benzenamine, 4-chloro-3-methoxy, pubchem3659, acmc-1chua
COC1=C(Cl)C=CC(N)=C1
157.60
157.6
13726-14-2
Characteristic
MFCD00672967
UN2233
LNKBDFVSILQKSI-UHFFFAOYSA-N
4-chloro-3-methoxyaniline
13103692
98%

Chemical Identifiers

13726-14-2
157.60
LNKBDFVSILQKSI-UHFFFAOYSA-N
13103692
COC1=C(Cl)C=CC(N)=C1
C7H8ClNO
MFCD00672967
3-methoxy-4-chloroaniline, 4-chloro-3-methoxyphenylamine, 4-chloro-m-anisidine, 2-chloro-5-aminoanisole, 5-amino-2-chloroanisole, 4-chloro-3-methoxy aniline, 4-chloro-3-methoxy-phenylamine, benzenamine, 4-chloro-3-methoxy, pubchem3659, acmc-1chua
4-chloro-3-methoxyaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROANISIDINES

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only