missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-methoxyaniline, 98%, Thermo Scientific Chemicals

$238.27 - $718.19

Chemical Identifiers

CAS 13726-14-2
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.60
MDL Number MFCD00672967
InChI Key LNKBDFVSILQKSI-UHFFFAOYSA-N
Synonym 3-methoxy-4-chloroaniline, 4-chloro-3-methoxyphenylamine, 4-chloro-m-anisidine, 2-chloro-5-aminoanisole, 5-amino-2-chloroanisole, 4-chloro-3-methoxy aniline, 4-chloro-3-methoxy-phenylamine, benzenamine, 4-chloro-3-methoxy, pubchem3659, acmc-1chua
PubChem CID 13103692
IUPAC Name 4-chloro-3-methoxyaniline
SMILES COC1=C(Cl)C=CC(N)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2833106
View Documents
Thermo Scientific Chemicals
H2833106
5 g
Each for $238.27
 
AAH2833114
View Documents
Thermo Scientific Chemicals
H2833114
25 g
Each for $718.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13726-14-2
157.60
LNKBDFVSILQKSI-UHFFFAOYSA-N
13103692
COC1=C(Cl)C=CC(N)=C1
C7H8ClNO
MFCD00672967
3-methoxy-4-chloroaniline, 4-chloro-3-methoxyphenylamine, 4-chloro-m-anisidine, 2-chloro-5-aminoanisole, 5-amino-2-chloroanisole, 4-chloro-3-methoxy aniline, 4-chloro-3-methoxy-phenylamine, benzenamine, 4-chloro-3-methoxy, pubchem3659, acmc-1chua
4-chloro-3-methoxyaniline

Specifications

13726-14-2
Characteristic
MFCD00672967
UN2233
LNKBDFVSILQKSI-UHFFFAOYSA-N
4-chloro-3-methoxyaniline
13103692
98%
78°C to 81°C
C7H8ClNO
5 g
3-methoxy-4-chloroaniline, 4-chloro-3-methoxyphenylamine, 4-chloro-m-anisidine, 2-chloro-5-aminoanisole, 5-amino-2-chloroanisole, 4-chloro-3-methoxy aniline, 4-chloro-3-methoxy-phenylamine, benzenamine, 4-chloro-3-methoxy, pubchem3659, acmc-1chua
COC1=C(Cl)C=CC(N)=C1
157.60
157.6
4-Chloro-3-methoxyaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROANISIDINES

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only