missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-3-nitrobenzophenone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2345114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chloro-3-nitrobenzophenone | |
104°C to 106°C | |
C13H8ClNO3 | |
25 g | |
YBDBYPQFIMSFJW-UHFFFAOYSA-N | |
(4-chloro-3-nitrophenyl)-phenylmethanone | |
41743 | |
99% |
56107-02-9 | |
233°C to 235°C (12 mmHg) | |
MFCD00007081 | |
4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb | |
[O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1 | |
261.66 | |
261.66 |
Chemical Identifiers
56107-02-9 | |
261.66 | |
YBDBYPQFIMSFJW-UHFFFAOYSA-N | |
41743 | |
[O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1 |
C13H8ClNO3 | |
MFCD00007081 | |
4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb | |
(4-chloro-3-nitrophenyl)-phenylmethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
H312+H332
EINECSNumber : 259-995-7
RTECSNumber : DJ0425000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only