missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-nitrobenzophenone, 99%, Thermo Scientific Chemicals

$55.33 - $134.59

Chemical Identifiers

CAS 56107-02-9
Molecular Formula C13H8ClNO3
Molecular Weight (g/mol) 261.66
MDL Number MFCD00007081
InChI Key YBDBYPQFIMSFJW-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb
PubChem CID 41743
IUPAC Name (4-chloro-3-nitrophenyl)-phenylmethanone
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2345114
View Documents
Thermo Scientific Chemicals
B2345114
25 g
Each for $55.33
 
AAB2345122
View Documents
Thermo Scientific Chemicals
B2345122
100 g
Each for $134.59
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56107-02-9
261.66
YBDBYPQFIMSFJW-UHFFFAOYSA-N
41743
[O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1
C13H8ClNO3
MFCD00007081
4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb
(4-chloro-3-nitrophenyl)-phenylmethanone

Specifications

56107-02-9
233°C to 235°C (12 mmHg)
MFCD00007081
4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb
[O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1
261.66
261.66
4-Chloro-3-nitrobenzophenone
104°C to 106°C
C13H8ClNO3
25 g
YBDBYPQFIMSFJW-UHFFFAOYSA-N
(4-chloro-3-nitrophenyl)-phenylmethanone
41743
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c

H312+H332

EINECSNumber : 259-995-7

RTECSNumber : DJ0425000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only