missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-nitrobenzophenone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2345122

 View more versions of this product

Catalog No. AAB2345122


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chloro-3-nitrobenzophenone
104°C to 106°C
C13H8ClNO3
100 g
YBDBYPQFIMSFJW-UHFFFAOYSA-N
(4-chloro-3-nitrophenyl)-phenylmethanone
41743
99%
56107-02-9
233°C to 235°C (12 mmHg)
MFCD00007081
4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb
[O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1
261.66
261.66

Chemical Identifiers

56107-02-9
261.66
YBDBYPQFIMSFJW-UHFFFAOYSA-N
41743
[O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1
C13H8ClNO3
MFCD00007081
4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb
(4-chloro-3-nitrophenyl)-phenylmethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c

H312+H332

EINECSNumber : 259-995-7

RTECSNumber : DJ0425000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only