missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Chlorosulfonyl)benzoic acid, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 405060050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(Chlorosulfonyl)benzoic acid | |
230.0°C | |
Authentic | |
Glass bottle | |
CO2HC6H4SO2Cl | |
4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811 | |
PTCSSXYPZOFISK-UHFFFAOYSA-N | |
4-chlorosulfonylbenzoic acid | |
82385 | |
96% |
10130-89-9 | |
White to Beige | |
96% | |
C7H5ClO4S | |
5 g | |
Solubility in water: decomposes. | |
C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl | |
220.63 | |
220.63 | |
Powder |
Chemical Identifiers
10130-89-9 | |
220.63 | |
4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811 | |
4-chlorosulfonylbenzoic acid |
C7H5ClO4S | |
PTCSSXYPZOFISK-UHFFFAOYSA-N | |
82385 | |
C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 233-367-2
RUO – Research Use Only