missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorothiobenzamide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1202118

 View more versions of this product

Catalog No. AAA1202118


Explore available promotions

Description

Description

4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

4-Chlorothiobenzamide
2521-24-6
C7H6ClNS
606603
Insoluble in water.
NC(=S)C1=CC=C(Cl)C=C1
171.64
171.65
123°C to 129°C
50 g
MFCD00040956
4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione
OKPUICCJRDBRJT-UHFFFAOYSA-N
4-chlorobenzene-1-carbothioamide
2734826
97%

Chemical Identifiers

2521-24-6
171.64
OKPUICCJRDBRJT-UHFFFAOYSA-N
2734826
C7H6ClNS
MFCD00040956
4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione
NC(=S)C1=CC=C(Cl)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only