Learn More
4-Chlorothiobenzamide, 97%, Thermo Scientific Chemicals
$96.96 - $496.73
Chemical Identifiers
CAS | 2521-24-6 |
---|---|
Molecular Formula | C7H6ClNS |
Molecular Weight (g/mol) | 171.64 |
MDL Number | MFCD00040956 |
InChI Key | OKPUICCJRDBRJT-UHFFFAOYSA-N |
Synonym | 4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione |
PubChem CID | 2734826 |
SMILES | NC(=S)C1=CC=C(Cl)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1202109
|
Thermo Scientific Chemicals
A1202109 |
10 g |
Each for $96.96
|
|
||||||
AAA1202114
|
Thermo Scientific Chemicals
A1202114 |
25 g |
Each for $176.97
|
|
||||||
AAA1202118
|
Thermo Scientific Chemicals
A1202118 |
50 g |
Each for $290.36
|
|
||||||
AAA1202122
|
Thermo Scientific Chemicals
A1202122 |
100 g |
Each for $496.73
|
|
||||||
Description
4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep away from oxidizing agents.
Chemical Identifiers
2521-24-6 | |
171.64 | |
OKPUICCJRDBRJT-UHFFFAOYSA-N | |
2734826 |
C7H6ClNS | |
MFCD00040956 | |
4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione | |
NC(=S)C1=CC=C(Cl)C=C1 |
Specifications
123°C to 129°C | |
10 g | |
MFCD00040956 | |
4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione | |
OKPUICCJRDBRJT-UHFFFAOYSA-N | |
4-chlorobenzenecarbothioamide | |
2734826 | |
97% |
2521-24-6 | |
C7H6ClNS | |
606603 | |
Insoluble in water. | |
NC(=S)C1=CC=C(Cl)C=C1 | |
171.64 | |
171.65 | |
4-Chlorothiobenzamide |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only