missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Cholesten-3-one, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1174314
Description
4-Cholesten-3-one is used as a chiral building block for proteomics research. It is an intestinal metabolite of cholesterol and exhibits an anti-obesity effect on animals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cholesten-3-one is used as a chiral building block for proteomics research. It is an intestinal metabolite of cholesterol and exhibits an anti-obesity effect on animals.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
4-Cholesten-3-one | |
79°C to 81°C | |
MFCD00003663 | |
4707774 | |
Slightly soluble in water. | |
+93° (c=1 in chloroform) | |
(8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one | |
91477 | |
384.65 |
601-57-0 | |
C27H44O | |
25 g | |
4-cholesten-3-one, cholest-4-en-3-one, cholestenone, 3-oxocholest-4-ene, delta sup 4-cholestenone, +-4-cholesten-3-one, unii-7t94nhd99c, 3-oxo-4-cholestene, 3-keto-4-cholestene, cholestenone delta 4 | |
NYOXRYYXRWJDKP-GYKMGIIDSA-N | |
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C | |
384.648 | |
CHEBI:16175 | |
≥98% |
Chemical Identifiers
601-57-0 | |
384.648 | |
NYOXRYYXRWJDKP-GYKMGIIDSA-N | |
91477 | |
(8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
C27H44O | |
MFCD00003663 | |
4-cholesten-3-one, cholest-4-en-3-one, cholestenone, 3-oxocholest-4-ene, delta sup 4-cholestenone, +-4-cholesten-3-one, unii-7t94nhd99c, 3-oxo-4-cholestene, 3-keto-4-cholestene, cholestenone delta 4 | |
CHEBI:16175 | |
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C |
Safety and Handling
EINECSNumber : 210-005-1
RTECSNumber : FZ7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only