missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Cholesten-3-one, 98+%, Thermo Scientific Chemicals
$147.25 - $522.67
Chemical Identifiers
CAS | 601-57-0 |
---|---|
Molecular Formula | C27H44O |
Molecular Weight (g/mol) | 384.648 |
MDL Number | MFCD00003663 |
InChI Key | NYOXRYYXRWJDKP-GYKMGIIDSA-N |
Synonym | 4-cholesten-3-one, cholest-4-en-3-one, cholestenone, 3-oxocholest-4-ene, delta sup 4-cholestenone, +-4-cholesten-3-one, unii-7t94nhd99c, 3-oxo-4-cholestene, 3-keto-4-cholestene, cholestenone delta 4 |
PubChem CID | 91477 |
ChEBI | CHEBI:16175 |
IUPAC Name | (8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
SMILES | CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1174306
|
Thermo Scientific Chemicals
A1174306 |
5 g |
Each for $147.25
|
|
|||||
AAA1174314
|
Thermo Scientific Chemicals
A1174314 |
25 g |
Each for $522.67
|
|
|||||
Description
4-Cholesten-3-one is used as a chiral building block for proteomics research. It is an intestinal metabolite of cholesterol and exhibits an anti-obesity effect on animals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cholesten-3-one is used as a chiral building block for proteomics research. It is an intestinal metabolite of cholesterol and exhibits an anti-obesity effect on animals.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
601-57-0 | |
384.648 | |
NYOXRYYXRWJDKP-GYKMGIIDSA-N | |
91477 | |
(8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
C27H44O | |
MFCD00003663 | |
4-cholesten-3-one, cholest-4-en-3-one, cholestenone, 3-oxocholest-4-ene, delta sup 4-cholestenone, +-4-cholesten-3-one, unii-7t94nhd99c, 3-oxo-4-cholestene, 3-keto-4-cholestene, cholestenone delta 4 | |
CHEBI:16175 | |
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C |
Specifications
601-57-0 | |
C27H44O | |
5 g | |
4-cholesten-3-one, cholest-4-en-3-one, cholestenone, 3-oxocholest-4-ene, delta sup 4-cholestenone, +-4-cholesten-3-one, unii-7t94nhd99c, 3-oxo-4-cholestene, 3-keto-4-cholestene, cholestenone delta 4 | |
NYOXRYYXRWJDKP-GYKMGIIDSA-N | |
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C | |
384.648 | |
CHEBI:16175 | |
≥98% |
79°C to 81°C | |
MFCD00003663 | |
4707774 | |
Slightly soluble in water. | |
+93° (c=1 in chloroform) | |
(8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one | |
91477 | |
384.65 | |
4-Cholesten-3-one |
Safety and Handling
EINECSNumber : 210-005-1
RTECSNumber : FZ7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only