missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Cyanoacetanilide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1494114

 View more versions of this product

Catalog No. AAA1494114


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4'-Cyanoacetanilide
204°C to 206°C
MFCD00001814
n-4-cyanophenyl acetamide, 4-cyanoacetanilide, 4-acetamidobenzonitrile, acetamide, n-4-cyanophenyl, 4'-cyanoacetanilide, p-acetamidobenzonitirle, acetanilide, 4'-cyano, p-acetylamino-benzonitrile, p-cyanoacetanilide, p-acetaminobenzonitrile
CC(=O)NC1=CC=C(C=C1)C#N
160.18
160.18
35704-19-9
C9H8N2O
25 g
UFKRTEWFEYWIHD-UHFFFAOYSA-N
N-(4-cyanophenyl)acetamide
37256
98%

Chemical Identifiers

35704-19-9
160.18
UFKRTEWFEYWIHD-UHFFFAOYSA-N
37256
CC(=O)NC1=CC=C(C=C1)C#N
C9H8N2O
MFCD00001814
n-4-cyanophenyl acetamide, 4-cyanoacetanilide, 4-acetamidobenzonitrile, acetamide, n-4-cyanophenyl, 4'-cyanoacetanilide, p-acetamidobenzonitirle, acetanilide, 4'-cyano, p-acetylamino-benzonitrile, p-cyanoacetanilide, p-acetaminobenzonitrile
N-(4-cyanophenyl)acetamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 252-685-2

RTECSNumber : AE1846000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only