Learn More
4'-Cyanoacetanilide, 98%, Thermo Scientific Chemicals
$54.78 - $189.92
Chemical Identifiers
CAS | 35704-19-9 |
---|---|
Molecular Formula | C9H8N2O |
Molecular Weight (g/mol) | 160.18 |
MDL Number | MFCD00001814 |
InChI Key | UFKRTEWFEYWIHD-UHFFFAOYSA-N |
Synonym | n-4-cyanophenyl acetamide, 4-cyanoacetanilide, 4-acetamidobenzonitrile, acetamide, n-4-cyanophenyl, 4'-cyanoacetanilide, p-acetamidobenzonitirle, acetanilide, 4'-cyano, p-acetylamino-benzonitrile, p-cyanoacetanilide, p-acetaminobenzonitrile |
PubChem CID | 37256 |
IUPAC Name | N-(4-cyanophenyl)acetamide |
SMILES | CC(=O)NC1=CC=C(C=C1)C#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1494106
|
Thermo Scientific Chemicals
A1494106 |
5 g |
Each for $54.78
|
|
|||||
AAA1494114
|
Thermo Scientific Chemicals
A1494114 |
25 g |
Each for $189.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
35704-19-9 | |
160.18 | |
UFKRTEWFEYWIHD-UHFFFAOYSA-N | |
37256 | |
CC(=O)NC1=CC=C(C=C1)C#N |
C9H8N2O | |
MFCD00001814 | |
n-4-cyanophenyl acetamide, 4-cyanoacetanilide, 4-acetamidobenzonitrile, acetamide, n-4-cyanophenyl, 4'-cyanoacetanilide, p-acetamidobenzonitirle, acetanilide, 4'-cyano, p-acetylamino-benzonitrile, p-cyanoacetanilide, p-acetaminobenzonitrile | |
N-(4-cyanophenyl)acetamide |
Specifications
35704-19-9 | |
C9H8N2O | |
5 g | |
UFKRTEWFEYWIHD-UHFFFAOYSA-N | |
N-(4-cyanophenyl)acetamide | |
37256 | |
98% |
204°C to 206°C | |
MFCD00001814 | |
n-4-cyanophenyl acetamide, 4-cyanoacetanilide, 4-acetamidobenzonitrile, acetamide, n-4-cyanophenyl, 4'-cyanoacetanilide, p-acetamidobenzonitirle, acetanilide, 4'-cyano, p-acetylamino-benzonitrile, p-cyanoacetanilide, p-acetaminobenzonitrile | |
CC(=O)NC1=CC=C(C=C1)C#N | |
160.18 | |
160.18 | |
4'-Cyanoacetanilide |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 252-685-2
RTECSNumber : AE1846000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only