missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyclohexylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3361603

 View more versions of this product

Catalog No. AAH3361603


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Cyclohexylbenzenesulfonyl chloride
52°C to 54°C
MFCD00625713
UN3261
4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, acmc-1awu9, chloro 4-cyclohexylphenyl sulfone, 4-cyclohexylphenylsulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexyl-1-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyclohexyl
ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1
258.76
258.77
56354-57-5
C12H15ClO2S
1 g
Moisture sensitive
CREMYEDHKUWVTB-UHFFFAOYSA-N
4-cyclohexylbenzenesulfonyl chloride
4339442
97%

Chemical Identifiers

56354-57-5
258.76
CREMYEDHKUWVTB-UHFFFAOYSA-N
4339442
ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1
C12H15ClO2S
MFCD00625713
4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, acmc-1awu9, chloro 4-cyclohexylphenyl sulfone, 4-cyclohexylphenylsulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexyl-1-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyclohexyl
4-cyclohexylbenzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only