missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyclohexylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

$155.22 - $940.48

Chemical Identifiers

CAS 56354-57-5
Molecular Formula C12H15ClO2S
Molecular Weight (g/mol) 258.76
MDL Number MFCD00625713
InChI Key CREMYEDHKUWVTB-UHFFFAOYSA-N
Synonym 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, acmc-1awu9, chloro 4-cyclohexylphenyl sulfone, 4-cyclohexylphenylsulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexyl-1-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyclohexyl
PubChem CID 4339442
IUPAC Name 4-cyclohexylbenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3361603
View Documents
Thermo Scientific Chemicals
H3361603
1 g
Each for $155.22
 
AAH3361609
View Documents
Thermo Scientific Chemicals
H3361609
10 g
Each for $940.48
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56354-57-5
258.76
CREMYEDHKUWVTB-UHFFFAOYSA-N
4339442
ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1
C12H15ClO2S
MFCD00625713
4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, acmc-1awu9, chloro 4-cyclohexylphenyl sulfone, 4-cyclohexylphenylsulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexyl-1-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyclohexyl
4-cyclohexylbenzenesulfonyl chloride

Specifications

56354-57-5
C12H15ClO2S
1 g
Moisture sensitive
CREMYEDHKUWVTB-UHFFFAOYSA-N
4-cyclohexylbenzenesulfonyl chloride
4339442
97%
52°C to 54°C
MFCD00625713
UN3261
4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, acmc-1awu9, chloro 4-cyclohexylphenyl sulfone, 4-cyclohexylphenylsulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexyl-1-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyclohexyl
ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1
258.76
258.77
4-Cyclohexylbenzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only